Assay Details
| Gene Symbol: ACADVL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174494.2 | NP_776919.1 | 10 - 11 | 1189 | 61 | |
| GenBank mRNA | BC103104.1 | - | 11 - 12 | 1223 | 61 | |
| BT030546.1 | - | 11 - 12 | 1237 | 61 | ||
| U30817.1 | - | 10 - 11 | 1189 | 61 | ||
More Information
Set Membership: |
|

ACADVL
Genomic Map