Assay Details
| Gene Symbol: ATP6V0D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174505.3 | NP_776930.1 | 2 - 3 | 356 | 68 | |
| GenBank mRNA | BC102305.1 | - | 2 - 3 | 383 | 68 | |
| J04204.1 | - | 2 - 3 | 357 | 68 | ||
More Information
Set Membership: |
|

ATP6V0D1
Genomic Map