Assay Details
| Gene Symbol: CHRNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174516.2 | NP_776941.1 | 8 - 9 | 1077 | 64 | |
| GenBank mRNA | BC147876.1 | - | 8 - 9 | 1112 | 64 | |
| X00962.1 | - | 1077 | 64 | |||
More Information
Set Membership: |
|

CHRNB1
Genomic Map