Assay Details
| Gene Symbol: DNASE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174534.2 | NP_776959.1 | 2 - 3 | 265 | 57 | |
| GenBank mRNA | AF311922.2 | - | 1 - 2 | 115 | 57 | |
| AF528510.1 | - | 1 - 2 | 108 | 57 | ||
| AJ001538.1 | - | 2 - 3 | 265 | 57 | ||
| BC142349.1 | - | 2 - 3 | 266 | 57 | ||
More Information
Set Membership: |
|

DNASE1
Genomic Map