Assay Details
| Gene Symbol: GABRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174545.1 | NP_776970.1 | 9 - 10 | 1155 | 59 | |
| GenBank mRNA | M55563.1 | - | 9 - 10 | 1155 | 59 | |
More Information
Set Membership: |
|

GABRG2
Genomic Map