Assay Details
| Gene Symbol: PABPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174569.2 | NP_776994.1 | 5 - 6 | 2817 | 74 | |
| GenBank mRNA | BC133468.1 | - | 5 - 6 | 736 | 74 | |
| BC133551.1 | - | 6 - 7 | 869 | 74 | ||
| BC133558.1 | - | 5 - 6 | 727 | 74 | ||
| X89969.1 | - | 2817 | 74 | |||
More Information
Set Membership: |
|

PABPN1
Genomic Map