Assay Details
| Gene Symbol: SLC1A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174600.2 | NP_777025.1 | 2 - 3 | 326 | 63 | |
| GenBank mRNA | BC120125.1 | - | 2 - 3 | 382 | 63 | |
| D82056.1 | - | 2 - 3 | 326 | 63 | ||
More Information
Set Membership: |
|

SLC1A3
Genomic Map