Assay Details
| Gene Symbol: SLC5A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174607.1 | NP_777032.1 | 1 | 1953 | 88 | |
| GenBank mRNA | S72877.1 | - | 1 | 450 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC5A3| Gene Symbol: SLC5A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174607.1 | NP_777032.1 | 1 | 1953 | 88 | |
| GenBank mRNA | S72877.1 | - | 1 | 450 | 88 | |
Set Membership: |
|