Assay Details
| Gene Symbol: SLC7A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174613.2 | NP_777038.1 | 2 - 3 | 756 | 64 | |
| GenBank mRNA | AF174615.1 | - | 2 - 3 | 756 | 64 | |
| BC126651.1 | - | 2 - 3 | 759 | 64 | ||
More Information
Set Membership: |
|

SLC7A5
Genomic Map