Assay Details
| Gene Symbol: UBQLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174628.1 | NP_777053.1 | 8 - 9 | 1335 | 66 | |
| GenBank mRNA | AY035552.1 | - | 8 - 9 | 1335 | 66 | |
| BC120031.1 | - | 8 - 9 | 1559 | 66 | ||
More Information
Set Membership: |
|

UBQLN1
Genomic Map