Assay Details
| Gene Symbol: CYP11B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174638.3 | NP_777063.2 | 2 - 3 | 399 | 63 | |
| GenBank mRNA | AH003250.2 | - | 2 - 3 | 397 | 63 | |
| D00185.1 | - | 399 | 63 | |||
More Information
Set Membership: |
|

CYP11B1
Genomic Map