Assay Details
| Gene Symbol: PAFAH1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174664.2 | NP_777089.1 | 3 - 4 | 577 | 86 | |
| GenBank mRNA | AB098963.1 | - | 3 - 4 | 250 | 86 | |
| BC103452.1 | - | 3 - 4 | 263 | 86 | ||
| D49678.1 | - | 3 - 4 | 577 | 86 | ||
More Information
Set Membership: |
|

PAFAH1B2
Genomic Map