Assay Details
Gene Symbol: PAFAH1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174665.2 | NP_777090.1 | 3 - 4 | 241 | 82 | |
GenBank mRNA | BC120202.1 | - | 3 - 4 | 266 | 82 | |
D30789.1 | - | 3 - 4 | 241 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
