Assay Details
| Gene Symbol: GABARAPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174675.2 | NP_777100.1 | 3 - 4 | 336 | 70 | |
| GenBank mRNA | AF020262.1 | - | 3 - 4 | 336 | 70 | |
| AY117147.1 | - | 3 - 4 | 296 | 70 | ||
| BC102902.1 | - | 3 - 4 | 357 | 70 | ||
| BT020958.1 | - | 3 - 4 | 345 | 70 | ||
More Information
Set Membership: |
|

GABARAPL2
Genomic Map