Assay Details
| Gene Symbol: HSD17B14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174701.2 | NP_777126.1 | 4 - 5 | 348 | 60 | |
| GenBank mRNA | AF202997.1 | - | 4 - 5 | 348 | 60 | |
| AF238859.1 | - | 4 - 5 | 348 | 60 | ||
| BC133473.1 | - | 4 - 5 | 354 | 60 | ||
More Information
Set Membership: |
|

HSD17B14
Genomic Map