Assay Details
| Gene Symbol: MGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174707.3 | NP_777132.1 | 2 - 3 | 124 | 82 | |
| GenBank mRNA | AB098895.1 | - | 2 - 3 | 137 | 82 | |
| AF210379.1 | - | 2 - 3 | 124 | 82 | ||
| AY911361.1 | - | 2 - 3 | 148 | 82 | ||
| BC102587.1 | - | 2 - 3 | 154 | 82 | ||
| X07363.1 | - | 95 | 82 | |||
More Information
Set Membership: |
|

MGP
Genomic Map