Assay Details
| Gene Symbol: CPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174750.2 | NP_777175.1 | 6 - 7 | 719 | 55 | |
| GenBank mRNA | BC146042.1 | - | 6 - 7 | 720 | 55 | |
| M61851.1 | - | 6 - 7 | 719 | 55 | ||
| Z33906.1 | - | 719 | 55 | |||
More Information
Set Membership: |
|

CPA1
Genomic Map