Assay Details
| Gene Symbol: RPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174760.2 | NP_777185.1 | 1 - 2 | 54 | 68 | |
| GenBank mRNA | AB099008.1 | - | 1 | 49 | 69 | |
| AF143815.1 | - | 1 - 2 | 54 | 68 | ||
| AY911380.1 | - | 2 - 3 | 62 | 68 | ||
| BC104528.1 | - | 1 | 45 | 68 | ||
| BT021081.1 | - | 1 - 2 | 34 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC104528.1, NM_174760.2 transcripts.
Set Membership: |
|

RPL10
Genomic Map