Assay Details
| Gene Symbol: PIK3R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174796.1 | NP_777221.1 | 2 - 3 | 210 | 69 | |
| GenBank mRNA | AF036256.1 | - | 2 - 3 | 210 | 69 | |
More Information
Set Membership: |
|

PIK3R3
Genomic Map