Assay Details
| Gene Symbol: NDUFS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174819.3 | NP_777244.1 | 4 - 5 | 386 | 68 | |
| GenBank mRNA | M58469.1 | - | 4 - 5 | 386 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFS3| Gene Symbol: NDUFS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174819.3 | NP_777244.1 | 4 - 5 | 386 | 68 | |
| GenBank mRNA | M58469.1 | - | 4 - 5 | 386 | 68 | |
Set Membership: |
|