Assay Details
| Gene Symbol: BSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174842.2 | NP_777267.1 | 5 - 6 | 437 | 121 | |
| GenBank mRNA | AF057133.1 | - | 5 - 6 | 437 | 121 | |
More Information
Set Membership: |
|

BSP5
Genomic Map