Assay Details
| Gene Symbol: GUCY1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175778.2 | NP_786972.1 | 2 - 3 | 542 | 76 | |
| GenBank mRNA | KX258202.1 | - | 2 - 3 | 542 | 76 | |
| X54014.1 | - | 542 | 76 | |||
More Information
Set Membership: |
|

GUCY1A1
Genomic Map