Assay Details
| Gene Symbol: ATP5F1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175796.3 | NP_786990.1 | 4 - 5 | 623 | 135 | |
| GenBank mRNA | BC116099.1 | - | 4 - 5 | 612 | 135 | |
| DQ403115.1 | - | 1 - 2 | 67 | 135 | ||
| M20929.1 | - | 4 - 5 | 623 | 135 | ||
More Information
Set Membership: |
|

ATP5F1B
Genomic Map