Assay Details
| Gene Symbol: ATP5F1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175796.3 | NP_786990.1 | 5 - 6 | 813 | 111 | |
| GenBank mRNA | BC116099.1 | - | 5 - 6 | 802 | 111 | |
| DQ403115.1 | - | 2 - 3 | 257 | 111 | ||
| M20929.1 | - | 5 - 6 | 813 | 111 | ||
| X05605.1 | - | 281 | 111 | |||
More Information
Set Membership: |
|

ATP5F1B
Genomic Map