Assay Details
| Gene Symbol: ATP6AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175806.2 | NP_787000.1 | 2 - 3 | 307 | 63 | |
| GenBank mRNA | BC104511.1 | - | 2 - 3 | 275 | 63 | |
| U10039.1 | - | 2 - 3 | 307 | 63 | ||
More Information
Set Membership: |
|

ATP6AP1
Genomic Map