Assay Details
| Gene Symbol: ALDH6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175811.2 | NP_787005.1 | 7 - 8 | 900 | 68 | |
| GenBank mRNA | L08643.1 | - | 7 - 8 | 900 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALDH6A1| Gene Symbol: ALDH6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175811.2 | NP_787005.1 | 7 - 8 | 900 | 68 | |
| GenBank mRNA | L08643.1 | - | 7 - 8 | 900 | 68 | |
Set Membership: |
|