Assay Details
| Gene Symbol: NDUFA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176658.2 | NP_788831.1 | 3 - 4 | 261 | 92 | |
| GenBank mRNA | BC146193.1 | - | 3 - 4 | 259 | 92 | |
| X68585.1 | - | 261 | 92 | |||
More Information
Set Membership: |
|

NDUFA7
Genomic Map