Assay Details
| Gene Symbol: SSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176665.2 | NP_788838.1 | 1 - 2 | 74 | 65 | |
| GenBank mRNA | BC103234.1 | - | 2 - 3 | 128 | 65 | |
| X13698.1 | - | 74 | 65 | |||
More Information
Set Membership: |
|

SSB
Genomic Map