Assay Details
| Gene Symbol: ATP5F1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176670.2 | NP_788843.1 | 3 - 4 | 432 | 68 | |
| GenBank mRNA | BC146208.1 | - | 3 - 4 | 394 | 68 | |
| X17019.1 | - | 432 | 68 | |||
More Information
Set Membership: |
|

ATP5F1D
Genomic Map