Assay Details
| Gene Symbol: FUT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176851.1 | NP_789821.1 | 1 - 2 | 92 | 94 | |
| GenBank mRNA | AJ132773.2 | - | 1 - 2 | 92 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ132773.2, NM_176851.1 transcripts.
Set Membership: |
|

FUT6
Genomic Map