Assay Details
| Gene Symbol: B4GALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177512.2 | NP_803478.1 | 5 - 6 | 1263 | 110 | |
| GenBank mRNA | AF515786.2 | - | 5 - 6 | 861 | 110 | |
| BC120415.1 | - | 5 - 6 | 1232 | 110 | ||
| M13214.1 | - | 5 - 6 | 878 | 110 | ||
| X14558.1 | - | 1263 | 110 | |||
More Information
Set Membership: |
|

B4GALT1
Genomic Map