Assay Details
| Gene Symbol: GLYAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177513.2 | NP_803479.1 | 2 - 3 | 207 | 97 | |
| GenBank mRNA | AF045032.1 | - | 2 - 3 | 207 | 97 | |
| AJ223301.1 | - | 2 - 3 | 207 | 97 | ||
| BC112536.1 | - | 2 - 3 | 227 | 97 | ||
More Information
Set Membership: |
|

GLYAT
Genomic Map