Assay Details
| Gene Symbol: GSTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177516.1 | NP_803482.1 | 1 - 2 | 74 | 69 | |
| GenBank mRNA | BC102704.1 | - | 1 - 2 | 48 | 69 | |
| X61233.1 | - | 74 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102704.1, NM_177516.1, X61233.1 transcripts.
Set Membership: |
|

GSTP1
Genomic Map