Assay Details
| Gene Symbol: GSTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177516.1 | NP_803482.1 | 3 - 4 | 215 | 116 | |
| GenBank mRNA | BC102704.1 | - | 3 - 4 | 189 | 116 | |
| X61233.1 | - | 215 | 116 | |||
More Information
Set Membership: |
|

GSTP1
Genomic Map