Assay Details
| Gene Symbol: GSTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177516.1 | NP_803482.1 | 6 - 7 | 513 | 83 | |
| GenBank mRNA | BC102704.1 | - | 6 - 7 | 487 | 83 | |
| X61233.1 | - | 513 | 83 | |||
More Information
Set Membership: |
|

GSTP1
Genomic Map