Assay Details
| Gene Symbol: CYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180994.2 | NP_851337.1 | 2 - 3 | 235 | 97 | |
| GenBank mRNA | AF421161.1 | - | 2 - 3 | 196 | 97 | |
| AF421162.1 | - | 2 - 3 | 196 | 97 | ||
| AF421163.1 | - | 2 - 3 | 196 | 97 | ||
| E05472.1 | - | 316 | 97 | |||
| EF541122.1 | - | 2 - 3 | 210 | 97 | ||
| FJ768675.1 | - | 2 - 3 | 213 | 97 | ||
| J00002.1 | - | 2 - 3 | 230 | 97 | ||
| J00003.1 | - | 2 - 3 | 235 | 97 | ||
More Information
Set Membership: |
|

CYM
Genomic Map