Assay Details
| Gene Symbol: CYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180994.2 | NP_851337.1 | 3 - 4 | 357 | 80 | |
| GenBank mRNA | E05472.1 | - | 438 | 80 | ||
| EF541122.1 | - | 3 - 4 | 332 | 80 | ||
| FJ768675.1 | - | 3 - 4 | 335 | 80 | ||
| J00002.1 | - | 3 - 4 | 352 | 80 | ||
| J00003.1 | - | 3 - 4 | 357 | 80 | ||
| J00004.1 | - | 3 - 4 | 316 | 80 | ||
More Information
Set Membership: |
|

CYM
Genomic Map