Assay Details
| Gene Symbol: CYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180994.2 | NP_851337.1 | 6 - 7 | 787 | 76 | |
| GenBank mRNA | AF421161.1 | - | 6 - 7 | 748 | 76 | |
| AF421164.1 | - | 4 - 5 | 511 | 76 | ||
| AF421165.1 | - | 4 - 5 | 511 | 76 | ||
| EF541122.1 | - | 6 - 7 | 762 | 76 | ||
| FJ768675.1 | - | 6 - 7 | 765 | 76 | ||
| J00002.1 | - | 6 - 7 | 782 | 76 | ||
| J00003.1 | - | 6 - 7 | 787 | 76 | ||
More Information
Set Membership: |
|

CYM
Genomic Map