Assay Details
| Gene Symbol: CSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181007.2 | NP_851350.2 | 2 - 3 | 397 | 76 | |
| GenBank mRNA | EE252986.1 | - | 2 - 3 | 418 | 76 | |
| EV793172.1 | - | 2 - 3 | 337 | 76 | ||
| M33268.1 | - | 2 - 3 | 339 | 76 | ||
| M65218.1 | - | 1 - 2 | 116 | 76 | ||
| M80327.1 | - | 1 - 2 | 117 | 76 | ||
More Information
Set Membership: |
|

CSH2
Genomic Map