Assay Details
| Gene Symbol: CSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164321.1 | NP_001157793.1 | 3 - 4 | 577 | 106 | |
| NM_181007.2 | NP_851350.2 | 4 - 5 | 685 | 106 | ||
| GenBank mRNA | AB098813.1 | - | 460 | 106 | ||
| BC149388.1 | - | 3 - 4 | 503 | 106 | ||
| CX736016.1 | - | 1 - 2 | 163 | 106 | ||
| EE252986.1 | - | 4 - 5 | 706 | 106 | ||
| EV793172.1 | - | 4 - 5 | 625 | 106 | ||
| J02840.1 | - | 4 - 5 | 608 | 106 | ||
| M33268.1 | - | 4 - 5 | 627 | 106 | ||
| M65219.1 | - | 2 - 3 | 288 | 106 | ||
| M80328.1 | - | 2 - 3 | 290 | 106 | ||
More Information
Set Membership: |
|

CSH2
Genomic Map