Assay Details
| Gene Symbol: GHRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181020.3 | NP_851363.1 | 3 - 4 | 372 | 62 | |
| GenBank mRNA | AB022596.1 | - | 3 - 4 | 331 | 62 | |
| AB022597.1 | - | 3 - 4 | 383 | 62 | ||
| AF184896.1 | - | 3 - 4 | 292 | 62 | ||
| AY008835.1 | - | 3 - 4 | 327 | 62 | ||
More Information
Set Membership: |
|

GHRHR
Genomic Map