Assay Details
| Gene Symbol: PPARG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181024.2 | NP_851367.1 | 6 - 7 | 1451 | 85 | |
| GenBank mRNA | AY179866.1 | - | 6 - 7 | 1292 | 85 | |
| Y12419.1 | - | 1347 | 85 | |||
| Y12420.1 | - | 1451 | 85 | |||
More Information
Set Membership: |
|

PPARG
Genomic Map