Assay Details
| Gene Symbol: CRABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181028.2 | NP_851371.1 | 2 - 3 | 535 | 108 | |
| GenBank mRNA | AY821681.1 | - | 2 - 3 | 255 | 108 | |
| BC112728.1 | - | 2 - 3 | 374 | 108 | ||
| M17253.1 | - | 2 - 3 | 535 | 108 | ||
| M36808.1 | - | 2 - 3 | 348 | 108 | ||
| X07436.1 | - | 325 | 108 | |||
More Information
Set Membership: |
|

CRABP1
Genomic Map