Assay Details
| Gene Symbol: CRABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181028.2 | NP_851371.1 | 3 - 4 | 655 | 111 | |
| GenBank mRNA | AY821681.1 | - | 3 - 4 | 372 | 108 | |
| BC112728.1 | - | 3 - 4 | 491 | 108 | ||
| M17253.1 | - | 3 - 4 | 655 | 111 | ||
| M36808.1 | - | 3 - 4 | 465 | 108 | ||
| X07436.1 | - | 442 | 108 | |||
More Information
Set Membership: |
|

CRABP1
Genomic Map