Assay Details
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203358.2 | NP_976234.2 | 1 - 2 | 178 | 88 | |
GenBank mRNA | DV884152.1 | - | 1 - 2 | 303 | 88 | |
DV906486.1 | - | 1 - 2 | 135 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_203358.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
