Assay Details
| Gene Symbol: REG3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205787.1 | NP_991356.1 | 1 - 2 | 108 | 66 | |
| GenBank mRNA | BC134799.1 | - | 2 - 3 | 130 | 66 | |
| M59794.1 | - | 1 - 2 | 108 | 66 | ||
More Information
Set Membership: |
|

REG3A
Genomic Map