Assay Details
| Gene Symbol: REG3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205787.1 | NP_991356.1 | 2 - 3 | 229 | 79 | |
| GenBank mRNA | BC134799.1 | - | 3 - 4 | 251 | 79 | |
| M59794.1 | - | 2 - 3 | 229 | 79 | ||
More Information
Set Membership: |
|

REG3A
Genomic Map