Assay Details
| Gene Symbol: REG3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205787.1 | NP_991356.1 | 4 - 5 | 494 | 83 | |
| GenBank mRNA | M59794.1 | - | 4 - 5 | 494 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

REG3A| Gene Symbol: REG3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205787.1 | NP_991356.1 | 4 - 5 | 494 | 83 | |
| GenBank mRNA | M59794.1 | - | 4 - 5 | 494 | 83 | |
Set Membership: |
|