Assay Details
| Gene Symbol: FAHD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205790.1 | NP_991359.1 | 4 - 5 | 696 | 65 | |
| GenBank mRNA | AY192436.1 | - | 4 - 5 | 696 | 65 | |
| BC112705.1 | - | 4 - 5 | 686 | 65 | ||
More Information
Set Membership: |
|

FAHD2A
Genomic Map