Assay Details
| Gene Symbol: FYTTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001137.1 | NP_001001137.1 | 3 - 4 | 400 | 136 | |
| GenBank mRNA | AJ344095.1 | - | 3 - 4 | 400 | 136 | |
| BC118174.1 | - | 136 | ||||
More Information
Set Membership: |
|

FYTTD1
Genomic Map